Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,3,4-Tri-O-acetyl-1-thio-beta-L-fucopyranoside 98.0+%, TCI America™
SDP

Catalog No. p-7132781 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 84635-54-1
Molecular Formula C13H20O7S
Molecular Weight (g/mol) 320.356
MDL Number MFCD00080803
InChI Key JOLBZRHXWOHZNS-MCNNAKBESA-N
PubChem CID 14131721
IUPAC Name [(2S,3R,4R,5S,6R)-4,5-diacetyloxy-2-methyl-6-methylsulfanyloxan-3-yl] acetate
SMILES CC1C(C(C(C(O1)SC)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 1 g
Formula Weight 320.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,3,4-Tri-O-acetyl-1-thio-beta-L-fucopyranoside
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.