Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl 2-fluorophenylacetate, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Specifications
Specifications
Color | Colorless |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4890 to 1.4910 |
Quantity | 5 g |
Formula Weight | 168.06 |
Percent Purity | 99% |
Physical Form | Liquid |
Chemical Name or Material | Methyl 2-fluorophenylacetate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction