Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Furancarboxylate 99.0+%, TCI America™
SDP

Catalog No. p-7129483 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
250 mL

Chemical Identifiers

CAS 611-13-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00003236
InChI Key HDJLSECJEQSPKW-UHFFFAOYSA-N
Synonym methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester
PubChem CID 11902
IUPAC Name methyl furan-2-carboxylate
SMILES COC(=O)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 181°C
Quantity 25 mL
UN Number 2810
Formula Weight 126.11
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Furancarboxylate
EINECSNumber (5)-0046
RTECSNumber LV1950000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.