Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Aminopyrazine-2-carboxylate 98.0+%, TCI America™
SDP

Catalog No. p-7117297 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 16298-03-6
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00010097
InChI Key INCSQLZZXBPATR-UHFFFAOYSA-N
Synonym methyl 3-amino-2-pyrazinecarboxylate, methyl 2-aminopyrazine-3-carboxylate, 3-aminopyrazine-2-carboxylic acid methyl ester, methyl 3-aminopyrazinecarboxylate, methyl 3-amino-2-pyrazine carboxylate, 3-amino-pyrazine-2-carboxylic acid methyl ester, 3-amino-2-pyrazinecarboxylic acid methyl ester, pyrazinecarboxylic acid, 3-amino-, methyl ester, methyl-3-aminopyrazin-2-carboxylat, zlchem 76
PubChem CID 72669
IUPAC Name methyl 3-aminopyrazine-2-carboxylate
SMILES COC(=O)C1=NC=CN=C1N

Specifications

Melting Point 172°C
Quantity 1 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Aminopyrazine-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.