Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Methoxysalicylate 98.0+%, TCI America™
SDP

Catalog No. p-7133067 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 6342-70-7
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00239511
InChI Key BWRCJLJJIXYLNV-UHFFFAOYSA-N
Synonym 3-Methoxysalicylic Acid Methyl Ester, Methyl 2-Hydroxy-3-methoxybenzoate, 2-Hydroxy-3-methoxybenzoic Acid Methyl Ester, Methyl 2-Hydroxy-m-anisate, 2-Hydroxy-m-anisic Acid Methyl Ester
PubChem CID 80651
IUPAC Name methyl 2-hydroxy-3-methoxybenzoate
SMILES COC1=CC=CC(=C1O)C(=O)OC

Specifications

Melting Point 68°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Methoxysalicylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.