Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride, Thermo Scientific™

Catalog No. MO00998DE
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 180258-45-1
Molecular Formula C7H11ClN2O2
Molecular Weight (g/mol) 190.63
MDL Number MFCD01318761
InChI Key HEOKCJUUKIPIMM-UHFFFAOYSA-N
Synonym methyl 4-amino-1-methyl-1h-pyrrole-2-carboxylate hydrochloride, methyl 4-amino-1-methylpyrrole-2-carboxylate hydrochloride, methyl-4-amino-1-methyl pyrrole-2-carboxylate hydrochloride, 4-amino-1-methyl-1h-pyrrole-2-carboxylic acid-methyl ester hcl, 4-amino-1-methylpyrrole-2-carboxylic acid methyl ester hydrochloride, h-nh 4-mepyl-2-ome hcl, methyl 4-amino-1-methyl-pyrrole-2-carboxylate hydrochloride, methyl-4-amino-1-methyl pyrrole-2-carboxylate, hcl, n-methyl-2-carbomethoxy-4-aminopyrrole hydrochloride, 3-amino-1-methyl-5-methyloxycarbonylpyrrol hydrochloride
PubChem CID 2794656
SMILES Cl.COC(=O)C1=CC(N)=CN1C

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 5 g
IUPAC Name methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride
Formula Weight 190.63
Percent Purity 95%
Chemical Name or Material Methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.