Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-iodobutyrate, 95%, stabilized, Thermo Scientific Chemicals

Catalog No. p-4089808
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14273-85-9
Molecular Formula C5H9IO2
Molecular Weight (g/mol) 228.02
MDL Number MFCD00674066
InChI Key NBCIIVXSBPDKOM-UHFFFAOYSA-N
Synonym methyl 4-iodobutyrate, butanoic acid, 4-iodo-, methyl ester, butyric acid, 4-iodo-, methyl ester, methyl-4-iodobutyrate, acmc-1bwlv, methyl gamma-iodobutyrate, 4-iodobutyric acid methyl ester, butanoic acid, 4-iodo-,methyl ester
PubChem CID 547991
IUPAC Name methyl 4-iodobutanoate
SMILES COC(=O)CCCI

Specifications

Color Colorless to Yellow
Density 1.6890g/mL
Boiling Point 80°C to 83°C (11.0 mmHg)
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula I(CH2)3COOCH3
Quantity 1 g
Specific Gravity 1.689
Stabilizer Copper
Formula Weight 228.02
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 4-iodobutyrate, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.