Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Methoxyacetoacetate 97.0+%, TCI America™
SDP

Catalog No. p-7132501 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 41051-15-4
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00010183
InChI Key QGBPKJFJAVDUNC-UHFFFAOYSA-N
Synonym methyl 4-methoxyacetoacetate, methyl-4-methoxyacetoacetate, methyl 4-methoxy-3-oxobutyrate, butanoic acid, 4-methoxy-3-oxo-, methyl ester, methyl methoxyacetoacetate, 4-methoxyacetoacetic acid methyl ester, 4-methoxy-3-oxo-butyric acid methyl ester, pubchem10473, acmc-1aoxb, methyl4-methoxyacetoacetate
PubChem CID 123500
IUPAC Name methyl 4-methoxy-3-oxobutanoate
SMILES COCC(=O)CC(=O)OC

Specifications

Color Yellow
Boiling Point 113°C
Quantity 5 g
Formula Weight 146.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Methoxyacetoacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.