Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Methoxysalicylate 99.0+%, TCI America™
SDP

Catalog No. p-7132858 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5446-02-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00008424
InChI Key ZICRWXFGZCVTBZ-UHFFFAOYSA-N
Synonym methyl 4-methoxysalicylate, 2-hydroxy-4-methoxy-benzoic acid methyl ester, methyl-4-methoxysalicylate, benzoic acid, 2-hydroxy-4-methoxy-, methyl ester, 2-hydroxy-4-methoxybenzoic acid methyl ester, 4-methoxysalicylic acid methyl ester, acmc-2097hp, methyl 2-hydroxy-p-anisate, ksc494a7p, 2-methoxycarbonyl-5-methoxyphenol
PubChem CID 79523
IUPAC Name methyl 2-hydroxy-4-methoxybenzoate
SMILES COC1=CC(=C(C=C1)C(=O)OC)O

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 134°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Methoxysalicylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.