Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl cis-11-Octadecenoate 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
100 mg
500 mg
Specifications
Specifications
Color | Yellow |
Boiling Point | 136°C |
Quantity | 100 mg |
Formula Weight | 296.50 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | Methyl cis-11-Octadecenoate |
Safety and Handling
EINECSNumber | (2)-0798 |
TSCA | No |
Recommended Storage | Freezer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction