Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cyclopentanecarboxylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7077355
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4630-80-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00019290
InChI Key IIHIJFJSXPDTNO-UHFFFAOYSA-N
Synonym cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl
PubChem CID 78365
IUPAC Name methyl cyclopentanecarboxylate
SMILES COC(=O)C1CCCC1

Specifications

Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 128.17
Percent Purity 97%
Chemical Name or Material Methyl cyclopentanecarboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Store in a well-ventilated place. Keep container tightly closed.
EINECSNumber 225-049-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.