Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl iso-Butyl Ketone, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Specifications
Specifications
Formula Weight | 100.16 |
Physical Form | Liquid |
Grade | Meets ACS reagent chemical requirements |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 0.80kg |
Flash Point | 14.00°C |
Boiling Point | 116.5°C |
Quantity | 500mL |
Chemical Name or Material | Methyl iso-Butyl Ketone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction