Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Isobutyl Ketone ACS AR, Macron Fine Chemicals™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Glass
Steel Pail
Specifications
Specifications
Quantity | 500 mL |
Packaging | Glass |
Formula Weight | 100.16 |
Grade | ACS Reagent |
Chemical Name or Material | Methyl Isobutyl Ketone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction