Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Methacrylate, Stabilized with hydroquinone, BAKER™, J.T. Baker™

Catalog No. 02003484 Shop All Avantor Products
Click to view available options
Quantity:
4L
Packaging:
Narrow Mouth Amber Glass Bottle

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 80-62-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC

Specifications

Flash Point 10.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 4L
Specific Gravity 1L = 0.94kg
Formula Weight 100.12
Chemical Name or Material Methyl Methacrylate, Stabilized with hydroquinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.