Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Pentafluoropropionate 98.0+%, TCI America™
SDP

Catalog No. p-7135093 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 378-75-6
Molecular Formula C4H3F5O2
Molecular Weight (g/mol) 178.058
MDL Number MFCD00000430
InChI Key JMKJCPUVEMZGEC-UHFFFAOYSA-N
Synonym methyl pentafluoropropionate, methyl perfluoropropionate, methylpentafluoropropionate, pentafluoropropionic acid methyl ester, propanoic acid, pentafluoro-, methyl ester, methyl pentafluoropropanoate, perfluoropropionic acid, methyl ester, pentafluoropropanoic acid methyl ester, 2,2,3,3,3-pentafluoro-propionic acid methyl ester, methylester kyseliny pentafluorpropionove
PubChem CID 9783
IUPAC Name methyl 2,2,3,3,3-pentafluoropropanoate
SMILES COC(=O)C(C(F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 60°C
Quantity 5 g
UN Number 3272
Formula Weight 178.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Pentafluoropropionate
RTECSNumber UF6475500
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.