Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(-)-3-Bromoisobutyrate 97.0+%, TCI America™
SDP

Catalog No. p-7119505 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 98190-85-3
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00064487
InChI Key FKWNAVCXZSQYTA-UHFFFAOYNA-N
Synonym (S)-(-)-3-Bromo-2-methylpropionic Acid Methyl Ester, (S)-(-)-3-Bromoisobutyric Acid Methyl Ester, Methyl (S)-(-)-3-Bromo-2-methylpropionate
PubChem CID 5479124
IUPAC Name methyl 3-bromo-2-methylpropanoate
SMILES COC(=O)C(C)CBr

Specifications

Color Yellow
Quantity 1 g
Formula Weight 181.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-3-Bromoisobutyrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.