Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl tert-Butyl Ether, ULTRA RESI-ANALYZED™, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For use in Gas Chromatography (GC), Organic Residue Analysis; For Laboratory, Research, or Manufacturing Use
Specifications
Specifications
Flash Point | −27.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 1L |
Specific Gravity | 1L = 0.74kg |
Formula Weight | 88.15 |
Grade | ULTRA RESI-ANALYZED™ |
Chemical Name or Material | Methyl tert-Butyl Ether |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction