Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. p-4228840
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4160-61-6
Molecular Formula C7H10O3S
Molecular Weight (g/mol) 174.22
InChI Key MCUXKFHPGMEIIW-UHFFFAOYSA-N
Synonym methyl 4-oxotetrahydro-2h-thiopyran-3-carboxylate, methyl tetrahydro-4-oxo-2h-thiopyran-3-carboxylate, methyl 4-oxotetrahydrothiopyran-3-carboxylate, 2h-thiopyran-3-carboxylic acid, tetrahydro-4-oxo-, methyl ester, methyl4-oxotetrahydro-2h-thiopyran-3-carboxylate, methyl-4-oxotetrahydro-2h-thiopyran-3-carboxylate, 2,3,5,6-tetrahydro-3-carbomethoxythiopyran-4-one, 4-oxotetrahydro-2h-thiopyran-3-carbonsauremethylester, 4-oxotetrahydro-2h-thiopyran-3-carboxylic acid methyl ester, 4-oxo-tetrahydro-thiopyran-3-carboxylic acid methyl ester
PubChem CID 10374916
IUPAC Name methyl 4-oxothiane-3-carboxylate
SMILES COC(=O)C1CSCCC1=O

Specifications

Color Colorless to Yellow
Boiling Point 134°C (10.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 174.22
Percent Purity 95%
Physical Form Liquid or Solid
Chemical Name or Material Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.