Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle2-(3-methoxyphenyl)acetate, 97%, Thermo Scientific™

Catalog No. BTB09252EB
Click to view available options
:

Specifications

CAS

18927-05-4

CAS Min %

96.5

CAS Max %

100.0

Assay Percent Range

97%

Molecular Formula

C10H12O3

Linear Formula

CH3OC6H4CH2C(O)OCH3

Quantity

25g

MDL Number

MFCD00017205

Synonym

methyl 2-3-methoxyphenyl acetate, methyl 3-methoxyphenylacetate, benzeneacetic acid, 3-methoxy-, methyl ester, acetic acid, m-methoxyphenyl-, methyl ester, methyl 3-methoxyphenyl acetate, 3-methoxy-phenyl-acetic acid methyl ester, acmc-1brer, methyl m-methoxyphenylacetate, methyl 3-methoxy-phenylacetate

InChI Key

BSVIOYCZTJRBDB-UHFFFAOYSA-N

SMILES

COC1=CC=CC(=C1)CC(=O)OC

IUPAC Name

methyl 2-(3-methoxyphenyl)acetate

Molecular Weight (g/mol)

180.203

PubChem CID

519609

Formula Weight

180.2

Percent Purity

97%

Chemical Name or Material

Methyle2-(3-methoxyphenyl)acetate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.