Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate, 97%, Thermo Scientific™

Catalog No. p-2705182
Click to view available options
Quantity:
1 g
10 g
25 g

Chemical Identifiers

CAS 175137-27-6
Molecular Formula C7H4ClF3N2O2
Molecular Weight (g/mol) 240.57
MDL Number MFCD00068146
InChI Key VLUBJYUOPNGBOQ-UHFFFAOYSA-N
Synonym methyl 2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, methyl 2-chloro-4-trifluoromethyl-5-pyrimidinecarboxylate, methylchlorotrifluoromethylpyrimidinecarboxylate, methyl-2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, acmc-20aifi, pubchem10174, 2-chloro-4-trifluoromethyl-5-carbomethoxypyrimidine, methyl 2-chloro-4-trifluoromethyl pyrimidine-5-ca, 2-chloro-5-methoxycarbonyl-4-trifluoromethylpyrimidine, 2-chloro-5-methoxycarbonyl-4-trifluoromethyl pyrimidine
PubChem CID 3696469
IUPAC Name methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
SMILES COC(=O)C1=CN=C(Cl)N=C1C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 240.57
Percent Purity 97%
Chemical Name or Material Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS P Statement Harmful if inhaled. Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. Harmful in contact with skin. Harmful if swallowed.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.