Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle3,4-diaminobenzoate, 97%, Thermo Scientific™

Catalog No. RDP00052DA
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

36692-49-6

Molecular Formula

C8H10N2O2

Molecular Weight (g/mol)

166.18

MDL Number

MFCD00017098

InChI Key

IOPLHGOSNCJOOO-UHFFFAOYSA-N

Synonym

3,4-diaminobenzoic acid methyl ester, methyl3,4-diaminobenzoate, methyl-3,4-diaminobenzoate, benzoic acid, 3,4-diamino-, methyl ester, 3,4-diamino-benzoic acid methyl ester, methyl 3,4-diaminobenzenecarboxylate, pubchem4610, acmc-1bn7s, methyl 3,4-diamino-benzoate, methyl-3,4-diamino-benzoate

PubChem CID

135524

IUPAC Name

methyl 3,4-diaminobenzoate

SMILES

COC(=O)C1=CC=C(N)C(N)=C1

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Packaging

Amber Glass Bottle

Linear Formula

CH3OC(O)C6H3(NH2)2

Quantity

1 g

Formula Weight

166.18

Percent Purity

97%

Chemical Name or Material

Methyl 3, 4-diaminobenzoate

Hazard Category

Warning

Hazard Statement

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

Precautionary Statement

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.