Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue, HARLECO™, MilliporeSigma™

Catalog No. M28423 Shop All MilliporeSigma Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

61-73-4

Molecular Formula

C16H18ClN3S

Molecular Weight (g/mol)

319.851

InChI Key

CXKWCBBOMKCUKX-UHFFFAOYSA-M

Synonym

Swiss Blue

PubChem CID

6099

ChEBI

CHEBI:6872

IUPAC Name

[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride

SMILES

CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]

Specifications

Linear Formula

C16H18N3S · HCl

Formula Weight

373.91

Grade

Certified Biological Stain

Color

Blue

Quantity

500 g

Physical Form

Liquid

Chemical Name or Material

Methylene Blue, HARLECO™

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.