Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metyrapone, Tocris Bioscience™

Catalog No. 329250 Shop All R&D Systems Products
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

11-β hydroxylase inhibitor

Cytochrome P450 inhibitor. Blocks glucocorticoid synthesis via inhibition of steroid 11-β hydroxylase (CYP11B1) activity (IC50 = 7.83 μM). Also inhibits CYP3A4 and cytochrome P450-mediated ω/ω-1 hydroxylase activity.

Chemical Identifiers

CAS 54-36-4
Molecular Formula C14H14N2O
Molecular Weight (g/mol) 226.28
MDL Number MFCD00006397
InChI Key FJLBFSROUSIWMA-UHFFFAOYSA-N
Synonym metyrapone, metopirone, 2-methyl-1,2-di-3-pyridyl-1-propanone, methopyrapone, metopiron, methapyrapone, methopirapone, methopyrinine, methopyrone, metyrapon
PubChem CID 4174
ChEBI CHEBI:44241
IUPAC Name 2-methyl-1,2-bis(pyridin-3-yl)propan-1-one
SMILES CC(C)(C(=O)C1=CC=CN=C1)C1=CC=CN=C1

Specifications

Quantity 50 mg
Formula Weight 226.27
Percent Purity >99%
Chemical Name or Material Metyrapone
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.