Learn More
World Precision Instrument Meyhoefer Curette, 14cm, 1.5mm Diameter
SDP
Supplier:  World Precision Instrument 501773
Encompass_Preferred
The Meyhoefer curette has an ice cream scoop-shaped tip with semi-sharp edges designed for scraping. In ophthalmology, it may be used to scrape chalazion from the eyelids. It is perfect for incising cysts, large and small. This curette has a sturdy handle with a smooth surface that improves the grip. Its square shape give you a solid grip and helps alleviate finger strain.
Catalog No. 50-252-1403
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.