Learn More
Sklar Instruments Curette, Meyhoefer Chalazion Curette, Sklar 5 in., #2, 2mm Cup, Straight, Stainless Steel, EA.
Supplier: Sklar Instruments 653172
Curette, Meyhoefer Chalazion Curette, Sklar, 5 in., #2, 2mm Cup, Straight, Stainless Steel, EA. Sklar's Meyhoefer Chalazion Curette is an ophthalmic instrument used for removing chalazion. A small cyst formed by inflamation of a gland in the margin of the eyelid, a chalazion is similar to a sty but often requires an incision and drainage to be properly treated. The round, sharp scooping tip of the Meyhoefer curette is available in multiple sizes to treat various size cysts. Following an incision, the curette is used to scoop out the cyst. This product is the #2 curette and has a round 2 mm tip. Lifetime warranty against all manufacturer defects.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.