Learn More
Cytiva (Formerly Pall Lab) MicroFunnel™ Filter Funnel with Polycarbonate Membrane, 300 mL, 0.4 m, Sterile, Individually Bagged (20/pkg)
Ideal for Concentrating Organisms in Fluid Samples-Flat, smooth surface allows for easy elution of organisms or particulate.-Collection of organisms or particulate on one planar surface is ideal prior to electron microscopy (SEM) or light microscope (LM) techniques.-Ready-to-use, disposable design eliminates sample cross contamination. This product is not approved for use in medical, clinical, surgical or other patient applications.