Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MI 2 (MALT1 inhibitor), Tocris Bioscience™

Catalog No. p-4817123 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MALT1 inhibitor

MALT1 inhibitor (IC50 = 5.84 μM). Binds directly to MALT1 and irreversibly suppresses protease function. Decreases NF-κB activity induced by MALT1. Inhibits cell proliferation and MALT1-mediated cleavage activity. Suppresses human TMD8 and HBL-1 activated B cell-like diffuse large B cell lymphoma (ABC-DLBCL) tumor xenografts in mice and primary human ABC-DLBCLs ex vivo.

Chemical Identifiers

CAS 1047953-91-2
Molecular Formula C19H17Cl3N4O3
Molecular Weight (g/mol) 455.72
InChI Key TWJGQZBSEMDPQP-UHFFFAOYSA-N
Synonym mi 2 malt1 inhibitor, mi 2, 2-chloro-n-4-5-3,4-dichlorophenyl-3-2-methoxyethoxy-1h-1,2,4-triazol-1-yl phenyl acetamide, 2-chloro-n-4-5-3,4-dichlorophenyl-3-2-methoxyethoxy-1,2,4-triazol-1-yl phenyl acetamide, malt1 inhibitor mi-2, mi-2 malt1 inhibitor, mi 2 malt1 inhibitor; mi 2
PubChem CID 45942672
IUPAC Name 2-chloro-N-[4-[5-(3,4-dichlorophenyl)-3-(2-methoxyethoxy)-1,2,4-triazol-1-yl]phenyl]acetamide
SMILES COCCOC1=NN(C(=N1)C2=CC(=C(C=C2)Cl)Cl)C3=CC=C(C=C3)NC(=O)CCl

Specifications

Quantity 10 mg
Formula Weight 455.72
Percent Purity >98%
Chemical Name or Material MI 2 (MALT1 inhibitor)
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.