Learn More
NEOGEN MI BROTH 2ML AMPULES 50/PK
SDP
Supplier:  NEOGEN 700002411
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
For the detection of Total Coliforms and E. coli in water testing using the membrane filtration method.
Catalog No. NC9040070
missing translation for 'QuantityLeft'