Learn More
Ted Pella Inc MICA SHEETS 100/PK
SDP
Supplier:  Ted Pella Inc 54
Encompass
Sheet, Mica; PELCO; Grade V5; 10 x 40mm; .23mm T; Muscovite, die cut, ruby, flat, minimum of inclusions, minimum air and bubbles, very clean; For such EM applications as carbon filming and particle spraying
Catalog No. NC9655733
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.