Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Micro-Splitter Valves

Available in High and Low Pressure options

Supplier:  MilliporeSigma™ Supelco™ 54719U

 View more versions of this product

Catalog No. 11-100-4316


Description

Description

Specifications

Specifications

Max Pressure: 4000 psi
PEEK/PTFE
High Pressure
10-32 Fingertight Fittings
1/16 in. Tubing
4000 psi (Max)
3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.