Learn More
Electron Microscopy Sciences MAC Microanalysis Standard Aluminum Mount, 2, 3, or 5 mm
Supplier:  Electron Microscopy Sciences 80074AL
Encompass_Preferred
All of our standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. They are supplied with fully authenticated certificates of analyses and a location map for standard identification. Mounts are polished to 0.25 micron diamond and carbon coated. Supplied in 2, 3, or5 mm brass tube.
Type: Mounted
Size: 2, 3, or 5 mm
Catalog No. 50-266-88
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.