Learn More
Microbiologics™ Epower™ Candida albicans ATCC™ 10231™†
SDP
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
$1079.00
Specifications
ATCC® 10231™*
Biosafety Level 1
Form Lyophilized Pellet
Quantity 10/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
Catalog Number Mfr. No. Mean Assay Value Price Quantity & Availability  
12-111-242
SDP
View Documents
Microbiologics™
0443E3CRM
1.0-9.9 + 3 CFU/pellet
Each for $1,079.00
Add to Cart
12-111-243
SDP
View Documents
Microbiologics™
0443E7CRM
1.0-9.9 + 7 CFU/pellet
Each for $1,079.00
Add to Cart
Description
Description
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 10231™†
Specifications
Specifications
10231™*
1
Lyophilized Pellet
10/Pk.
Videos
SDS
Documents
Documents
Product Certifications
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.