Learn More
Microbiologics™ Cryptosporidium species Parasite Suspension
Shop All Microbiologics Products

- Ready-to-use
- In-Vitro Diagnostic (IVD) device that mimics patient sample
- Shelf-life: two years
- Medical IVD
- With Certificate of Performance
- Packaged in 1mL vial
QC of diagnostic kits and methods, microscopic examinations, acid-fast staining procedures, proficiency programs and educational purposes for parasitology
Compliance
CE marked
Specifications
Certifications/Compliance
CE
Content And Storage
Room Temperature
Form
Liquid
Media Type
Suspension
Product Type
Parasite Suspension
For Use With (Application)
QC of diagnostic kits and method, microscopic examinations, Acid fast staining procedures, proficiency programs and educational purposes for parasitology
Quantity
1 mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.