Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Yarrowia lipolytica ATCC™ 9773™†
SDP

For culture purposes and QC applications

$211.00 - $364.00

Specifications

Species Yarrowia lipolytica
Biosafety Level 1
Form Pellets
For Use With (Application) For culture purposes and QC applications.
Includes Peel-off identification label
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
23-024701
SDP
View Documents
Microbiologics™
0950P
KWIK-STIK 2/Pk.
Pack of 2 for $211.00
missing translation for 'QuantityLeft'
Add to Cart
 
23-016-401
SDP
View Documents
Microbiologics™
0950K
KWIK-STIK 6/Pk.
Pack of 6 for $364.00
missing translation for 'QuantityLeft'
Add to Cart
 
23-016-900
SDP
View Documents
Microbiologics™
0950L
Lyfo-Disk QC Control 6/Pk.
Pack of 6 for $362.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single Yarrowia lipolytica strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Yarrowia lipolytica strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 9773™†
  • †The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Specifications

Specifications

Yarrowia lipolytica
Pellets
Peel-off identification label
1
For culture purposes and QC applications.
Re-sealable Vial
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.