Learn More
Eppendorf™ Microcentrifuge Tubes™, Forensic DNA Grade

Ideal for working with medium–sized sample volumes. Eppendorf Tubes™, Forensic DNA Grade enables simple and safe processing of sample volumes up to 5mL.
$123.30 - $142.80
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
No. per Pack | 200 Pk. |
Packaging | 200 Tubes (4 Bags x 50 Tubes) |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Closure Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Type | Price | Quantity & Availability | |||||
E0030119606
![]() |
Eppendorf™
0030119606 |
Snap Cap |
Each for $123.30
|
|
|||||
E0030122402
|
Eppendorf™
0030122402 |
Screw Cap |
Each for $142.80
|
|
|||||
Description
- Conical bottom allows usage in existing adapters and racks
- Made from transparent polypropylene for easy visibility of pellets
- Free of plasticizers, biocides or mold release agents
- Sample evaporation during storage and incubation minimized by precise lid sealing
- Accidental lid opening from −86° to 80°C prevented by hinged lids
- Centrifugation stability of up to 25,000 x g prevents tube breakage
Applications
- Easier processing of samples up to 5mL
- Higher yields in DNA isolation, especially with samples with limited quantity of source material
- Convenient and safe cell culture applications
- Easier sample access lowers risk of contamination
- Preparation of master mixes and buffers
- Less storage space needed
- Safe cell and tissue lysis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Autoclavable | |
200 Pk. | |
Non-sterile |
Polypropylene | |
200 Tubes (4 Bags x 50 Tubes) | |
Cylindrical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.