Learn More
World Precision Instrument Microelectrode Holder (MPH6S)-MPH6S20
SDP
Supplier:  World Precision Instrument MPH6S20
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For use with these WPI Products: Piconozzle Kit (5430-XX)
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
Catalog No. 50-252-1064
missing translation for 'QuantityLeft'