Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Septa

MicroLiter septa for 9mm Screwthread Autosampler Vials are constructed with a wide array of chemical compatibilities to suit a variety of chromatography applications.

Supplier:  DWK Life Sciences 090021

 View more versions of this product

Catalog No. 15-111-034


Description

Description

Specifications

Specifications

Orange
FEP/Butyl
9 mm Autosampler Vials - MicroLiter
No
100
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.