Learn More
TWD Scientific LLC Micropette, Mechanical Pipette, Twdbiox, Single-Channel, Adjustable Volume In 0.1Ul Increments, Range 0.5 - 10Ul, 1/Case.
SDP
Supplier:  TWD Scientific LLC BX012101
Encompass_Preferred
Micropette Plus Mechanical Pipettes. Micropette Plus Features Single-Channel Adjustable Volume In 0.1Ul Increments Ranging From 0.5-10Ul. Price/Each
Catalog No. 50-228-5011
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.