Learn More
United Scientific™ Micropipette Tips, Low Retention, Bulk
SDPSCI_ED
Micropipette Tips, Low Retention, Bulk are disposable tips intended for use with most brands of pipettors.
Supplier:  United Scientific™ UNP10004
 View more versions of this product
Catalog No. S8538032
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • Deliver 10x lower retention levels with viscous samples, as compared to standard pipette tips
  • Highly homogenous surface is free of defects, preventing non-specific binding
  • Manufactured from ultra pure USP Class VI certified virgin polypropylene
  • Free of DNase, RNase and pyrogen
  • Natural color
  • Bulk packaging
Specifications
Specifications
Low Retention Micropipette Tips
Non-filtered
Natural
UNP10004
Pipetting
Bagged
1000 Pk.
Single-use
Polypropylene
200 μL
Non-sterile
Autoclavable
USP Class VI certified
Universal
UltraFine, Low Retention
Micropipette Tips
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.