Learn More
Electron Microscopy Sciences Sturkey Gold High Profile Disposible Microtome Blades, Standard Dispenser
Supplier: Electron Microscopy Sciences 6305921
GOLD disposable microtome blades are the ultimate blade for cutting all types of skin and hard tissue. It is a high-performance specialty blade and its durability and toughness make it the best solution for hard tissue.
Package: Standard Dispenser
Length: 80 mm
Angle: 35°
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.