Learn More
NEOGEN Milk Plate Count Agar, 500 g
Supplier: NEOGEN 700003308
Milk Plate Count Agar is used for the isolation and enumeration of microorganisms in milk and dairy products and is not intended for use in the diagnosis of disease or other conditions in humans.A medium recommended by the British Standards Institute and the International Organization for Standardization for the enumeration of viable bacteria in milk and other dairy products.For more information visit https://www.neogen.com/solutions/Microbiology/milk-plate-count-agar
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.