Learn More
Pluriselect MINI 70UM CYTOMETRY TUBES 100/
Supplier: Pluriselect 431007066
Bundle includes sterile pluriStrainers Mini with 70 µm mesh and sterile 5 ml polystyrene (PS) tubes (12x75mm). The combination of a flow cytometry tube and cell strainer offers a convenient way to prepare laboratory samples for flow cytometry analysis. A 70 µm polyethylenterephthalat (PET) mesh is incorporated into the housing of the pluriStrainer Mini 70 µm. The polystyrene round-bottom tubes are closed with a cap.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.