Learn More
Norgen Biotek Corp Plasmid MiniPrep Kit | 50 preps
Supplier: Norgen Biotek Corp 13300
High yield of plasmid DNA. Purify plasmids up to 13 kb. Plasmid DNA is ready for restriction digestion, bacterial transformation, sequencing and more. Rapid spin-column format and 96-well format available.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.