Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minocycline Hydrochloride, MP Biomedicals™

Catalog No. ICN15571883 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

13614-98-7

Molecular Formula

C23H28ClN3O7

Molecular Weight (g/mol)

493.94

MDL Number

MFCD00083669

InChI Key

KDLQIOPKJDNQIM-WUURTAMISA-N

Synonym

minocycline hydrochloride, minocycline hcl, minomycin, minocin, minocycline chloride, arestin, vectrin, minomax, periocline, tri-mino

PubChem CID

54685925

IUPAC Name

hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride

SMILES

[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

Color

Yellow

Quantity

250 mg

Formula Weight

493.9

Physical Form

Powder

Chemical Name or Material

Minocycline Hydrochloride

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.