Learn More
Medchemexpress LLC HY-108006 5mg Medchemexpress, Mirogabalin besylate CAS:1138245-21-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1080065MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-108006 5mg Mirogabalin besylate CAS:1138245-21-2 Mirogabalin besylate is a selective and orally available ligand for the α2δ subunit of voltage-gated calcium channels, with Kds of 13.5 nM, 22.7 nM, 27 nM, and 47.6 nM for human α2δ-1, human α2δ-2, rat α2δ-1, and rat α2δ-2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8498