Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ TransIT™-CHO Transfection Kit
Developed for high transfection efficiency and cell viability in CHO cell types. Does not require medium changes and can be performed in serum-containing medium.
Supplier: Mirus Bio™ MIR2176
Includes: TransIT-CHO reagent and CHO Mojo reagent, both supplied as ready-to-use solutions.
Description
- Unique Formulation: Each kit contains a base reagent and a booster reagent to optimize transfection performance in CHO cells
- Low Cellular Toxicity: Maintains cell density and reduces experimental bias
- High Efficiency Delivery: Allows 50 to 60% transfection efficiency in CHO-K1 cells
- Suitable for both transient and stable transfection
Specifications
Specifically optimized to provide exceptional transfection efficiency of plasmid DNA in CHO cells and cell types of associated lineage | |
Each | |
10mL 2.22mg/mL in 80% Ethanol, TransIT™-CHO reagent and 7mL 5mg/mL in 100% CHO Mojo reagent, both supplied as ready-to-use solution | |
Store at −20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction