Learn More
Hach Company Mixed-Parameter Standard, NIST, Wastewater, High Range
Supplier:  Hach Company LCA720
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Mixed-Parameter quality control standard for wastewater parameters, high range. Components of the standard solution (with the exception of ammonia) are traceable to SRM from NIST.COD – 500 mg/LPO4-P – 10.0 mg/LNH3-N – 25.0 mg/LNO3-N – 25.0 mg/LTN – 50.0 mg/LApplicable for TNTplus vial tests- TNT822 COD, 20-1500 mg/L O2- TNT825 Hg-free COD, 25-1000 mg/L O2- TNT845 Phosphate, 2-20 mg/L PO4-P- TNT832 Ammonia, 2-47 mg/L NH3-N- TNT836 Nitrate, 5-35 mg/L NO3-N- TNT828 Total Nitrogen, 20-100 mg/L TN
Catalog No. 50-199-7738
missing translation for 'QuantityLeft'