Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-MK 801 maleate, Tocris Bioscience™

Catalog No. p-4783070 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-competitive NMDA antagonist; acts at ion channel site

(+)-MK 801 maleate is a high affinity (Ki = 37.2 nM), selective and non-competitive NMDA receptor antagonist. (+)-MK 801 maleate acts by binding to a site located within the NMDA associated ion channel and thus prevents Ca2+ flux. It is an effective anti-ischemic agent in several animal models. (+)-MK 801 maleate increases motor activity in rat models. (+)-MK 801 maleate inhibits proliferation and increases apoptosis in hippocampal neural stem cells (NSCs). (+)-MK 801 maleate induces schizophrenia-like symptoms in rodents.

(-)-enantiomer also available.

Chemical Identifiers

CAS 77086-22-7
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.375
InChI Key QLTXKCWMEZIHBJ-PJGJYSAQSA-N
Synonym dizocilpine maleate, +-mk-801 hydrogen maleate, +-mk 801 maleate, dizocilpine hydrogen maleate, unii-6lr8c1b66q, dizocilpine maleate usan, dsstox_cid_25785, dsstox_rid_81126
PubChem CID 6420042
SMILES CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 337.37
Percent Purity >99%
Chemical Name or Material (+)-MK 801 maleate
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.