Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ML 145, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective antagonist for the GPR35 orphan receptor GPCR (IC50 = 20.1 nM). Over 1000-fold more selective for GPR35 compared to GPR55 antagonists.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 482.57 |
Percent Purity | >98% |
Chemical Name or Material | ML 145 |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction