Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ML 190, Tocris Bioscience™

Catalog No. p-4835948 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective κ opioid receptor antagonist

Selective κ opioid receptor (KOP) antagonist (IC50 = 120 nM in a β-arrestin assay); displays >267-fold selectivity over μ and δ opioid receptors.

Chemical Identifiers

CAS 1355244-02-8
Molecular Formula C27H32N6O3
Molecular Weight (g/mol) 488.592
InChI Key PMTIWRPLQBVEMR-UHFFFAOYSA-N
Synonym n-3-4-4-methoxyphenyl piperazin-1-yl propyl-2-1-methyl-6-oxopyrido 2,3-e pyrrolo 1,2-a pyrazin-5 6h-yl acetamide, n-3-4-4-methoxyphenyl-1-piperazinyl propyl-1-methyl-6-oxopyrido 2,3-e pyrrolo 1,2-a pyrazine-5 6h-acetamide, n-3-4-4-methoxyphenyl piperazin-1-yl propyl-2-13-methyl-7-oxo-2,8,10-triazatricyclo 7.4.0.0 2 ,? trideca-1 13 ,3,5,9,11-pentaen-8-yl acetamide
PubChem CID 44665680
SMILES CC1=C2C(=NC=C1)N(C(=O)C3=CC=CN32)CC(=O)NCCCN4CCN(CC4)C5=CC=C(C=C5)OC

Specifications

Quantity 10 mg
Formula Weight 488.58
Chemical Name or Material ML 190
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.